Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl phosphite, 98%, Acros Organics™

Click to view available options
Quantity:
1mL
100mL
122-52-1
C6H15O3P
166.157
MFCD00009084
BDZBKCUKTQZUTL-UHFFFAOYSA-N
phosphorous acid, triethyl ester,triethoxyphosphine,ethyl phosphite, eto 3p,triethylphosphite,phosphorous acid triethyl ester,fosforyn trojetylowy czech,unii-6b2r04s55g,hsdb 895,p oet 3,c2h5o 3p
31215
triethyl phosphite
CCOP(OCC)OCC
122-52-1
C6H15O3P
166.157
MFCD00009084
BDZBKCUKTQZUTL-UHFFFAOYSA-N
phosphorous acid, triethyl ester,triethoxyphosphine,ethyl phosphite, eto 3p,triethylphosphite,phosphorous acid triethyl ester,fosforyn trojetylowy czech,unii-6b2r04s55g,hsdb 895,p oet 3,c2h5o 3p
31215
triethyl phosphite
CCOP(OCC)OCC
Density 0.9650g/mL
Quantity 1mL
Assay Percent Range 98%
Linear Formula (C2H5O)3P
Beilstein 01, 330
Fieser 01,1212; 02,432; 03,304; 04,529; 05,693; 06,612; 07,387; 08,501; 11,555; 12,143
Merck Index 15, 9840
Formula Weight 166.16
Chemical Name or Material Triethyl phosphite, 97%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-552-5

RTECSNumber : TH1130000

TSCA : TSCA