Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylaluminum (15% in Hexane, ca. 1.0mol/L), TCI America™

Click to view available options
Quantity:
100mL
97-93-8
C6H15Al
114.168
MFCD00009015
VOITXYVAKOUIBA-UHFFFAOYSA-N
triethylaluminum,triethylaluminium,aluminum, triethyl,triethylalane,unii-h426e9h3tt,triethylaluminum solution, 1.0 m in hexanes,triethylaluminum solution, 25 wt. % in toluene,triethyl-aluminum,triethyl aluminium,triethyl-aluminium
16682930
triethylalumane
CC[Al](CC)CC
97-93-8
C6H15Al
114.168
MFCD00009015
VOITXYVAKOUIBA-UHFFFAOYSA-N
triethylaluminum,triethylaluminium,aluminum, triethyl,triethylalane,unii-h426e9h3tt,triethylaluminum solution, 1.0 m in hexanes,triethylaluminum solution, 25 wt. % in toluene,triethyl-aluminum,triethyl aluminium,triethyl-aluminium
16682930
triethylalumane
CC[Al](CC)CC
Color Colorless
Quantity 100mL
UN Number 3394
Formula Weight 114.17
Physical Form Clear Liquid at 20°C
Chemical Name or Material Triethylaluminum (15% in Hexane, ca. 1.0mol/L)

EINECSNumber : (2)-2227

RTECSNumber : BD2050000

TSCA : Yes