Alfa Aesar™ Triethylaluminum, 94%

Catalyst for polymerization of olefins, intermediate

Manufacturer: Alfa Aesar™ 4324314

Quantity 25g

 View more versions of this product

Catalog No. AA4324314

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 97-93-8
    Molecular Formula (C2H5)3Al
    Formula Weight 114.17
    MDL Number MFCD00009015
    UN Number UN3394
    Chemical Name or Material Triethylaluminum

    Specifications

    Formula Weight 114.17
    Health Hazard 2 GHS H Statement
    H225-H250-H260-H314
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Moisture Sensitive
    Physical Form Liquid
    CAS 97-93-8
    Odor Characteristic
    MDL Number MFCD00009015
    UN Number UN3394
    Density 0.835g/mL
    Melting Point −50°C
    Packaging Cylinder
    TSCA TSCA
    Boiling Point 128° to 130°C (50mmHg)
    Flash Point 18°C (−1°F)
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    Molecular Formula (C2H5)3Al
    Quantity 25g
    Health Hazard 1 Danger
    Chemical Name or Material Triethylaluminum
    Assay 94%

    GHS H Statement
    H225-H250-H260-H314
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit