Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

121-44-8
C6H15N
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
CHEBI:35026

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
CAS 121-44-8
Molecular Formula C6H15N
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILES CCN(CC)CC
IUPAC Name N,N-diethylethanamine
Molecular Weight (g/mol) 101.193
PubChem CID 8471
Packaging Narrow Mouth Amber Glass Bottle
ChEBI CHEBI:35026
Formula Weight 101.19
Specific Gravity 1L = 0.73kg
Flash Point −11.00°C
Chemical Name or Material Triethylamine
Grade BAKER ANALYZED™ Reagent
Show More Show Less