Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, Fisher BioReagents

Click to view available options
Quantity:
500mL
Packaging:
Glass Bottle
121-44-8
C6H15N
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane
8471
CHEBI:35026
N,N-diethylethanamine
CCN(CC)CC
121-44-8
C6H15N
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
triethylamine,ethanamine, n,n-diethyl,diethylamino ethane,triaethylamin,triethylamin,trietilamina,triethyl amine,n,n,n-triethylamine,net3,diethylaminoethane
8471
CHEBI:35026
N,N-diethylethanamine
CCN(CC)CC
Triethylamine is used as a solvent in automated synthesis protocols.
Boiling Point 90°C
Vapor Pressure 69mbar at 20°C
Refractive Index 1.397 to 1.403 (at 25°C)
Identification Pass Test
Melting Point -115°C
pH 12.4
Packaging Glass Bottle
Quantity 500mL
Color Colorless
Density 0.728g/cm³
Assay Percent Range ≥99 %
Linear Formula (C2H5)3N
Merck Index 15, 9831
Formula Weight 101.19
Physical Form Liquid
Percent Purity ≥99%
Grade Peptide Synthesis
Chemical Name or Material Triethylamine
Show More Show Less

DANGER!

Emergency Overview
Highly flammable. Harmful by inhalation, in contact with skin and if swallowed. May cause pulmonary edema. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:3
Instability:0

Recommended Storage : RT