Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine hydrobromide, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
636-70-4
C6H15N·HBr
182.1
MFCD00054287
NRTLTGGGUQIRRT-UHFFFAOYSA-N
triethylamine hydrobromide,ethanamine, n,n-diethyl-, hydrobromide,ethanamine, n,n-diethyl-, hydrobromide 1:1,n,n-diethylethanamine hydrobromide,acmc-20ajeg,et3n.hbr,triethylammonium bromide salt,triethylamine hydrobromide salt
2734074
N,N-diethylethanamine;hydrobromide
CCN(CC)CC.Br
636-70-4
C6H15N·HBr
182.1
MFCD00054287
NRTLTGGGUQIRRT-UHFFFAOYSA-N
triethylamine hydrobromide,ethanamine, n,n-diethyl-, hydrobromide,ethanamine, n,n-diethyl-, hydrobromide 1:1,n,n-diethylethanamine hydrobromide,acmc-20ajeg,et3n.hbr,triethylammonium bromide salt,triethylamine hydrobromide salt
2734074
N,N-diethylethanamine;hydrobromide
CCN(CC)CC.Br
Flash Point >65°C
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 100g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula (C2H5)3N·HBr
Formula Weight 182.1
Physical Form Crystalline Powder or Flakes
Percent Purity 99%
Chemical Name or Material Triethylamine hydrobromide, 99%
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISO

GHS Signal Word: Warning

EINECSNumber : 211-263-8

TSCA : TSCA