Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylammonium formate solution 2M (1:1, pH 6.5), Thermo Scientific™

Catalog Number AAH36155AE
Click to view available options
Quantity:
100 mL
585-29-5
C7H17NO2
147.218
MFCD00134943
PTMFUWGXPRYYMC-UHFFFAOYSA-N
unii-un4t038p8m, triethylamine formate, teaf, triethylammonium formate, triethylamine, formate, triethylammonium formate mi, triethylammoniumformate, triethylammonium format, hcooh et3n, formic acid triethylamine
11084032
N,N-diethylethanamine;formic acid
CCN(CC)CC.C(=O)O
585-29-5
C7H17NO2
147.218
MFCD00134943
PTMFUWGXPRYYMC-UHFFFAOYSA-N
unii-un4t038p8m, triethylamine formate, teaf, triethylammonium formate, triethylamine, formate, triethylammonium formate mi, triethylammoniumformate, triethylammonium format, hcooh et3n, formic acid triethylamine
11084032
N,N-diethylethanamine;formic acid
CCN(CC)CC.C(=O)O

 

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.01 g/mL
pH 6.5
Linear Formula (CH3CH2)3NH+ COO-
Quantity 100 mL
UN Number UN1760
Beilstein 4150472
Formula Weight 147.22
Concentration or Composition (by Analyte or Components) 2M (1:1)
Physical Form Liquid
Chemical Name or Material Triethylammonium formate solution
Show More Show Less
Hazard Category H302+H332-H314-H335
Hazard Statement GHS H Statement
H314-H302-H312-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit