Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Triethylammonium Formate Solution, Honeywell™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Plastic Bottle
585-29-5
C7H17NO2
147.218
MFCD00134943
PTMFUWGXPRYYMC-UHFFFAOYSA-N
11084032
N,N-diethylethanamine;formic acid
CCN(CC)CC.C(=O)O
585-29-5
C7H17NO2
147.218
MFCD00134943
PTMFUWGXPRYYMC-UHFFFAOYSA-N
11084032
N,N-diethylethanamine;formic acid
CCN(CC)CC.C(=O)O

volatile buffer, 1 M pH 6.0, for HPLC

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
For Use With Volatile buffer for HPLC
Linear Formula (CH3CH2)3NH+ HCOO-
UN Number NONH for all modes of transport
Beilstein 4150472
Formula Weight 147.22g/mol
Packaging Plastic Bottle
Density 1.00g/mL at 20°C
Physical State Liquid
pH 5.9 to 6.1 (at 25°C)
Color Colorless
Quantity 100mL
Chemical Name or Material Triethylammonium Formate Solution
Show More Show Less

P305 + P351 + P338

H315-H319