Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylborane, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass bottle
97-94-9, 109-99-9
C6H15B
98.00
MFCD00009022
LALRXNPLTWZJIJ-UHFFFAOYSA-N
triethylboron, borane, triethyl, triethylborine, boron triethyl, boron ethyl, unii-z3s980z4p3, hsdb 897, triethylboran, borethyl, triethyl borane
7357
triethylborane
CCB(CC)CC
97-94-9, 109-99-9
C6H15B
98.00
MFCD00009022
LALRXNPLTWZJIJ-UHFFFAOYSA-N
triethylboron, borane, triethyl, triethylborine, boron triethyl, boron ethyl, unii-z3s980z4p3, hsdb 897, triethylboran, borethyl, triethyl borane
7357
triethylborane
CCB(CC)CC
Lewis acid
Packaging AcroSeal™ Glass bottle
Color Amber to Colorless
Specific Gravity 0.86
Quantity 100mL
Density 0.8600g/mL
Flash Point −17°C
Linear Formula (C2H5)3B
Fieser 09,482; 10,416; 15,333; 16,160; 17,363
Merck Index 15, 9833
Formula Weight 98
Physical Form Liquid
Chemical Name or Material Triethylborane, 1M solution in THF
Show More Show Less

GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Suspect

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 202-620-9