Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylborane, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
97-94-9,109-99-9
C6H15B
97.996
MFCD00009022
LALRXNPLTWZJIJ-UHFFFAOYSA-N
triethylboron,borane, triethyl,triethylborine,boron triethyl,boron ethyl,unii-z3s980z4p3,hsdb 897,triethylboran,borethyl,triethyl borane
7357
triethylborane
B(CC)(CC)CC
97-94-9,109-99-9
C6H15B
97.996
MFCD00009022
LALRXNPLTWZJIJ-UHFFFAOYSA-N
triethylboron,borane, triethyl,triethylborine,boron triethyl,boron ethyl,unii-z3s980z4p3,hsdb 897,triethylboran,borethyl,triethyl borane
7357
triethylborane
B(CC)(CC)CC
Lewis acid
Specific Gravity 0.86
Packaging AcroSeal™ Glass Bottle
Quantity 800mL
Flash Point −17°C
Density 0.8600g/mL
Color Amber to Colorless
Linear Formula (C2H5)3B
Fieser 09,482; 10,416; 15,333; 16,160; 17,363
Merck Index 15, 9833
Formula Weight 98
Physical Form Liquid
Chemical Name or Material Triethylborane, 1M solution in THF
Show More Show Less

GHS H Statement
Highly flammable liquid and vapour. Harmful if swallowed. Harmful if inhaled. Causes severe skin burns and eye damage. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer if inhaled. Reacts violently with water. May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger