Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyle1,1,2-ethanetricarboxylate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
7459-46-3
C11H18O6
246.259
MFCD00009154
TVWZLLYAJDSSCJ-UHFFFAOYSA-N
triethyl 1,1,2-ethanetricarboxylate,1,1,2-ethanetricarboxylic acid, triethyl ester,2-ethoxycarbonyl-succinic acid diethyl ester,triethyl ethane tricarboxylate,1,1,2-triethyl ethane-1,1,2-tricarboxylate,triethyl ethane-1,2,2-tricarboxylate,ethane-1,1,2-tricarboxylic acid triethyl ester,ethane-1,1,2-tricarboxylic acid, triethyl ester,diethyl 2-ethoxycarbonyl butane-1,4-dioate,acmc-209ovd
81961
triethyl ethane-1,1,2-tricarboxylate
CCOC(=O)CC(C(=O)OCC)C(=O)OCC
7459-46-3
C11H18O6
246.259
MFCD00009154
TVWZLLYAJDSSCJ-UHFFFAOYSA-N
triethyl 1,1,2-ethanetricarboxylate,1,1,2-ethanetricarboxylic acid, triethyl ester,2-ethoxycarbonyl-succinic acid diethyl ester,triethyl ethane tricarboxylate,1,1,2-triethyl ethane-1,1,2-tricarboxylate,triethyl ethane-1,2,2-tricarboxylate,ethane-1,1,2-tricarboxylic acid triethyl ester,ethane-1,1,2-tricarboxylic acid, triethyl ester,diethyl 2-ethoxycarbonyl butane-1,4-dioate,acmc-209ovd
81961
triethyl ethane-1,1,2-tricarboxylate
CCOC(=O)CC(C(=O)OCC)C(=O)OCC
Quantity 100g
Infrared Spectrum Authentic
Color Colorless
Packaging Glass bottle
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Density 1.07g/mL
Assay Percent Range 99%
Linear Formula C2H5O2CCH2CH(CO2C2H5)2
Beilstein 02, 813
Formula Weight 246.26
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Triethyle1, 1, 2-ethanetricarboxylate
Show More Show Less