Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl 2-phosphonobutyrate, 98%, Acros Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
17145-91-4
C10H21O5P
252.25
MFCD00041347
GYUCVQSNZFRDRL-UHFFFAOYSA-N
triethyl 2-phosphonobutyrate,ethyl 2-diethoxyphosphoryl butanoate,ethyl 2-diethylphosphono butyrate,butanoic acid, 2-diethoxyphosphinyl-, ethyl ester,triethyl 2-phosphonobutanoate,.alpha.-diethylphosphono butanoic acid, ethyl ester,butyric acid, 2-phosphono-, triethyl ester,butyric acid, triethyl ester,triethyl 2-phosphonobu-tyrate,triethyl-2-phosphono butyrate
229053
ethyl 2-diethoxyphosphorylbutanoate
CCC(C(=O)OCC)P(=O)(OCC)OCC
17145-91-4
C10H21O5P
252.25
MFCD00041347
GYUCVQSNZFRDRL-UHFFFAOYSA-N
triethyl 2-phosphonobutyrate,ethyl 2-diethoxyphosphoryl butanoate,ethyl 2-diethylphosphono butyrate,butanoic acid, 2-diethoxyphosphinyl-, ethyl ester,triethyl 2-phosphonobutanoate,.alpha.-diethylphosphono butanoic acid, ethyl ester,butyric acid, 2-phosphono-, triethyl ester,butyric acid, triethyl ester,triethyl 2-phosphonobu-tyrate,triethyl-2-phosphono butyrate
229053
ethyl 2-diethoxyphosphorylbutanoate
CCC(C(=O)OCC)P(=O)(OCC)OCC
for C-C Bond Formation (Olefination)
Specific Gravity 1.064
Quantity 5mL
Color Colorless
Density 1.0640g/mL
Refractive Index 1.4315 to 1.4335
Packaging Glass bottle
Boiling Point 152.0°C to 154.0°C (14.0mmHg)
Infrared Spectrum Authentic
Flash Point >110°C
Assay Percent Range 97.5% min. (GC)
Linear Formula (O)P(CH3CH2O)2CH(CH2CH3)C(O)OCH2CH3
Formula Weight 252.25
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Triethyl 2-phosphonobutyrate, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning