Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol Dimethyl Ether, Stabilized, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
112-49-2
C8H18O4
178.23
MFCD00008504
YFNKIDBQEZZDLK-UHFFFAOYSA-N
triglyme,triethylene glycol dimethyl ether,2,5,8,11-tetraoxadodecane,1,2-bis 2-methoxyethoxy ethane,glyme 4,ansul ether 161,glyme-3,tegdime,tegdme,unii-32yxg88kk0
8189
CHEBI:44842
1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
COCCOCCOCCOC
112-49-2
C8H18O4
178.23
MFCD00008504
YFNKIDBQEZZDLK-UHFFFAOYSA-N
triglyme,triethylene glycol dimethyl ether,2,5,8,11-tetraoxadodecane,1,2-bis 2-methoxyethoxy ethane,glyme 4,ansul ether 161,glyme-3,tegdime,tegdme,unii-32yxg88kk0
8189
CHEBI:44842
1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
COCCOCCOCCOC
Refractive Index 1.4220 to 1.424
Boiling Point 216°C
Flash Point 113°C
Melting Point -45°C
Specific Gravity 0.986
Viscosity 2.5 mPa.s (20°C)
Acidity 0.05mg KOH/g max.
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1L
Density 0.9860g/mL
Color Undesignated
Water 0.1% max. (K.F.)
Stabilizer 0.01% BHT
Assay Percent Range 99%
Linear Formula CH3OCH2CH2OCH2CH2OCH2CH2OCH3
Merck Index 15, 9861
Solubility Information Solubility in water: soluble. Other solubilities: miscible with hydrocarbon solvents
Formula Weight 178.23
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Triethylene glycol dimethyl ether, Stabilized
Show More Show Less

GHS H Statement:
May damage fertility or the unborn child.
May form explosive peroxides.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger