Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylenetetramine, 60%, ACROS Organics™

Click to view available options
Quantity:
50g
500g
2.5kg
112-24-3
C6H18N4
146.24
MFCD00008169
VILCJCGEZXAXTO-UHFFFAOYSA-N
triethylenetetramine,trientine,trien,teta,tecza,triethylene tetramine,araldite hardener hy 951,1,2-ethanediamine, n,n'-bis 2-aminoethyl,deh 24,araldite hy 951
5565
CHEBI:39501
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
C(CNCCNCCN)N
112-24-3
C6H18N4
146.24
MFCD00008169
VILCJCGEZXAXTO-UHFFFAOYSA-N
triethylenetetramine,trientine,trien,teta,tecza,triethylene tetramine,araldite hardener hy 951,1,2-ethanediamine, n,n'-bis 2-aminoethyl,deh 24,araldite hy 951
5565
CHEBI:39501
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
C(CNCCNCCN)N
Specific Gravity 0.98
Boiling Point 280.0°C
Flash Point 129°C
Quantity 500g
Density 0.9800g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4960 to 1.5000
Color Undesignated
Melting Point -35.0°C
Assay Percent Range 55% min. (GC)
Linear Formula H2NCH2CH2NHCH2CH2NHCH2CH2NH2
Beilstein 04,255
Fieser 01,1204
Merck Index 15,9828
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol
Formula Weight 146.24
Physical Form Liquid
Percent Purity 60%
Grade Technical
Chemical Name or Material Triethylenetetramine, 60%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful in contact with skin.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 203-950-6

RTECSNumber : YE6650000

TSCA : TSCA