Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyloxonium tetrafluoroborate, 1.0M in dichloromethane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog Number AA42934AE
Click to view available options
Quantity:
100 mL
Packaging:
ChemSeal™ bottles
368-39-8
C6H15BF4O
189.99
MFCD00044423
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
2723982
triethyloxidanium;tetrafluoroborate
F[B-](F)(F)F.CC[O+](CC)CC
368-39-8
C6H15BF4O
189.99
MFCD00044423
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
2723982
triethyloxidanium;tetrafluoroborate
F[B-](F)(F)F.CC[O+](CC)CC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.328 g/mL
Odor Ether-like
Packaging ChemSeal™ bottles
Linear Formula (CH3CH2)3OBF4
Quantity 100 mL
UN Number UN2922
Beilstein 3598090
Sensitivity Moisture sensitive
Formula Weight 189.99
Concentration or Composition (by Analyte or Components) 1.0M in dichloromethane
Physical Form Liquid
Chemical Name or Material Triethyloxonium tetrafluoroborate
Show More Show Less
Hazard Category H314-H336-H350
Hazard Statement GHS H Statement
H314-H318-H351
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 206-705-1
RTECSNumber RR4584700
TSCA Yes
storageNote1 −30° to -10°C
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit