Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
368-39-8,75-09-2
C6H15BF4O
189.989
MFCD00044423
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
triethyloxonium tetrafluoroborate,triethyloxonium fluoroborate,triethyloxonium fluoborate,triethyloxonium borofluoride,triethoxonium fluoroborate,meerwein's reagent,triethyloxidanium tetrafluoroborate,unii-z0b19dd36j,triethyloxonium tetraflouroborate,boron tetrafluoride triethyl oxonium
2723982
triethyloxidanium;tetrafluoroborate
[B-](F)(F)(F)F.CC[O+](CC)CC
368-39-8,75-09-2
C6H15BF4O
189.989
MFCD00044423
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
triethyloxonium tetrafluoroborate,triethyloxonium fluoroborate,triethyloxonium fluoborate,triethyloxonium borofluoride,triethoxonium fluoroborate,meerwein's reagent,triethyloxidanium tetrafluoroborate,unii-z0b19dd36j,triethyloxonium tetraflouroborate,boron tetrafluoride triethyl oxonium
2723982
triethyloxidanium;tetrafluoroborate
[B-](F)(F)(F)F.CC[O+](CC)CC
Alkylation reagent
Linear Formula (C2H5)3OBF4
Fieser 01,1210; 02,430; 03,303; 04,527; 05,691; 06,611; 07,386; 08,500; 09,482; 10,417; 15,337
Density 1.3280g/mL
Formula Weight 189.99
Solubility Solubility in water: reacts.
Specific Gravity 1.328
Quantity 500mL
Chemical Name or Material Triethyloxonium tetrafluoroborate, 1M Solution in Methylene Chloride

GHS H Statement:
Suspected of causing cancer.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 206-705-1

TSCA : TSCA