Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triethylphosphine, 97%

Manufacturer:  Alfa Aesar™ 3017703

Quantity 1g

 View more versions of this product

Catalog No. AA3017703


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

554-70-1
118.16
UN2845
Triethylphosphine
(C2H5)3P
MFCD00009256
14,9675

Specifications

0.81g/mL
126° to 128°C
554-70-1
97%
118.16
UN2845
Triethylphosphine
Air & Moisture Sensitive
1g
Liquid
(C2H5)3P
MFCD00009256
14,9675
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H250-H252-H332
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Harmful if inhaled.

GHS P Statement
P210-P222-P261-P303+P361+P353-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit