Triethylphosphine, 97%

Manufacturer: Alfa Aesar 3017706

Quantity 5g

 View more versions of this product

Catalog No. AA3017706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 554-70-1
    Molecular Formula (C2H5)3P
    Formula Weight 118.16
    MDL Number MFCD00009256
    UN Number UN2845
    Merck Index 14,9675
    Chemical Name or Material Triethylphosphine

    Specifications

    Formula Weight 118.16
    Quantity 5g
    TSCA TSCA
    UN Number UN2845
    Merck Index 14,9675
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H250-H252-H332
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P210-P222-P261-P303+P361+P353-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Physical Form Liquid
    Density 0.81g/mL
    Molecular Formula (C2H5)3P
    CAS 554-70-1
    Assay Percent Range 97%
    MDL Number MFCD00009256
    Chemical Name or Material Triethylphosphine
    Assay 97%

    GHS H Statement
    H225-H250-H252-H332
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Harmful if inhaled.

    GHS P Statement
    P210-P222-P261-P303+P361+P353-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit