Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylphosphine oxide, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
597-50-2
C6H15OP
134.159
MFCD00013914
ZSSWXNPRLJLCDU-UHFFFAOYSA-N
triethylphosphine oxide,phosphine oxide, triethyl,triethylphoshorus oxide,c2h5 3p=o,triethylphospine oxide,et3po,triethylphosphino-1-one,triethyl oxo phosphorane,diethylphosphoryl ethane,triethylphosphine oxide 1g
79061
1-diethylphosphorylethane
CCP(=O)(CC)CC
597-50-2
C6H15OP
134.159
MFCD00013914
ZSSWXNPRLJLCDU-UHFFFAOYSA-N
triethylphosphine oxide,phosphine oxide, triethyl,triethylphoshorus oxide,c2h5 3p=o,triethylphospine oxide,et3po,triethylphosphino-1-one,triethyl oxo phosphorane,diethylphosphoryl ethane,triethylphosphine oxide 1g
79061
1-diethylphosphorylethane
CCP(=O)(CC)CC
Infrared Spectrum Authentic
Boiling Point 84.0°C to 85.0°C (3.0mmHg)
Quantity 1g
Packaging Glass bottle
Melting Point 52.0°C to 53.0°C
Color White
Assay Percent Range 98+%
Formula Weight 134.16
Physical Form Needles
Percent Purity ≥98%
Chemical Name or Material Triethylphosphine oxide
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger