Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylsilane, 98+%, Thermo Scientific Chemicals

Catalog Number aaa1032009
Click to view available options
Quantity:
10 g
50 g
250 g
617-86-7
C6H15Si
115.271
MFCD00009018
QXTIBZLKQPJVII-UHFFFAOYSA-N
triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
6327258
triethylsilicon
CC[Si](CC)CC
617-86-7
C6H15Si
115.271
MFCD00009018
QXTIBZLKQPJVII-UHFFFAOYSA-N
triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
6327258
triethylsilicon
CC[Si](CC)CC

Triethylsilane is used for the hydrosilation of olefins to prepare alkyl silanes. It acts as a reducing agent in the reduction of 2-chromanols, since it has an active hydride. It acts as a catalyst for redox initiated cationic polymerization, regioselective reductive coupling of enones and allenes, and Beckmann rearrangement of cyclododecanone oxime. It is associated with trifluoroacetic acid and involved in the selective reduction of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsilane is used for the hydrosilation of olefins to prepare alkyl silanes. It acts as a reducing agent in the reduction of 2-chromanols, since it has an active hydride. It acts as a catalyst for redox initiated cationic polymerization, regioselective reductive coupling of enones and allenes, and Beckmann rearrangement of cyclododecanone oxime. It is associated with trifluoroacetic acid and involved in the selective reduction of alkenes.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, water and bases.
TRUSTED_SUSTAINABILITY
Boiling Point 107°C to 108°C
Melting Point -157°C
Quantity 10 g
Linear Formula (CH3CH2)3SiH
UN Number UN1993
Beilstein 1098278
Flash Point −6°C (21°F)
Solubility Information Miscible with water.
Formula Weight 116.28
Percent Purity ≥98%
Refractive Index 1.412
Sensitivity Moisture sensitive
Density 0.728
Chemical Name or Material Triethylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-535-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit