Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylsilane, 99%, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
617-86-7
C6H16Si
116.28
QXTIBZLKQPJVII-UHFFFAOYSA-N
triethylsilane,hydride,silane, triethyl,triethylhydrosilane,triethylsilyl,silane e3h,c6h16si,triethylsilyl radical,silane, triethyl-,,triethylsilane tes
6327258
triethylsilicon
CC[Si](CC)CC
617-86-7
C6H16Si
116.28
QXTIBZLKQPJVII-UHFFFAOYSA-N
triethylsilane,hydride,silane, triethyl,triethylhydrosilane,triethylsilyl,silane e3h,c6h16si,triethylsilyl radical,silane, triethyl-,,triethylsilane tes
6327258
triethylsilicon
CC[Si](CC)CC
Flash Point -3°C
Boiling Point 107°C to 108°C
Refractive Index 1.4100 to 1.412
Specific Gravity 0.72
Packaging AcroSeal™ Glass Bottle
Density 0.7200g/mL
Quantity 1L
Assay Percent Range 98.5% min. (GC)
Linear Formula (C2H5)3SiH
Merck Index 15,9841
Solubility Information Solubility in water: decomposes
Formula Weight 116.28
Percent Purity 99%
Chemical Name or Material Triethylsilane
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 210-535-3