Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Triethylsilyl)acetylene, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
1777-03-3
C8H16Si
140.301
MFCD00075373
FWSPXZXVNVQHIF-UHFFFAOYSA-N
triethylsilyl acetylene,ethynyltriethylsilane,triethyl ethynyl silane,triethylsilylacetylene,silane, triethylethynyl,triethylsilyl acetylen,triethylsilyl-acetylene,triethyl-ethynyl-silane
637947
triethyl(ethynyl)silane
CC[Si](CC)(CC)C#C
1777-03-3
C8H16Si
140.301
MFCD00075373
FWSPXZXVNVQHIF-UHFFFAOYSA-N
triethylsilyl acetylene,ethynyltriethylsilane,triethyl ethynyl silane,triethylsilylacetylene,silane, triethylethynyl,triethylsilyl acetylen,triethylsilyl-acetylene,triethyl-ethynyl-silane
637947
triethyl(ethynyl)silane
CC[Si](CC)(CC)C#C
Quantity 1g
Density 0.783g/cm3
Assay Percent Range 97%
Formula Weight 140.3
Chemical Name or Material (Triethylsilyl)acetylene