Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Triethylsilyl)acetylene, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
1777-03-3
C8H16Si
140.3
MFCD00075373
FWSPXZXVNVQHIF-UHFFFAOYSA-N
triethylsilyl acetylene,ethynyltriethylsilane,triethyl ethynyl silane,triethylsilylacetylene,silane, triethylethynyl,triethylsilyl acetylen,triethylsilyl-acetylene,triethyl-ethynyl-silane
637947
triethyl(ethynyl)silane
CC[Si](CC)(CC)C#C
1777-03-3
C8H16Si
140.3
MFCD00075373
FWSPXZXVNVQHIF-UHFFFAOYSA-N
triethylsilyl acetylene,ethynyltriethylsilane,triethyl ethynyl silane,triethylsilylacetylene,silane, triethylethynyl,triethylsilyl acetylen,triethylsilyl-acetylene,triethyl-ethynyl-silane
637947
triethyl(ethynyl)silane
CC[Si](CC)(CC)C#C
Specific Gravity 0.783
Refractive Index 1.4310 to 1.4330
Density 0.7830g/mL
Quantity 5g
Packaging Glass bottle
Flash Point 17°C
Assay Percent Range 96% min. (GC)
Linear Formula (CH3CH2)3SiC≡CH
Beilstein 04,IV,393
Formula Weight 140.3
Percent Purity 97%
Chemical Name or Material (Triethylsilyl)acetylene
Show More Show Less

GHS Signal Word: Danger