Alfa Aesar™ Triethyltin bromide

Manufacturer: Alfa Aesar™ 7113514

Quantity 25g

 View more versions of this product

Catalog No. AA7113514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2767-54-6
    Molecular Formula (C2H5)3SnBr
    Formula Weight 285.79
    MDL Number MFCD00000052
    UN Number UN2788
    Synonym Bromotriethylstannane
    Chemical Name or Material Triethyltin bromide

    Specifications

    Formula Weight 285.79
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    CAS 2767-54-6
    Flash Point 99°C (210°F)
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Boiling Point 223° to 225°C
    Molecular Formula (C2H5)3SnBr
    MDL Number MFCD00000052
    UN Number UN2788
    Synonym Bromotriethylstannane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Density 1.63g/mL
    Refractive Index 1.520
    Sensitivity Moisture Sensitive
    Melting Point −13.5°C
    Quantity 25g
    Chemical Name or Material Triethyltin bromide

    GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit