Alfa Aesar™ Triethyltin chloride

Manufacturer: Alfa Aesar™ 3960114

Quantity 25g

 View more versions of this product

Catalog No. AA3960114

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 994-31-0
    Formula Weight 241.33
    MDL Number MFCD00040414
    UN Number UN2788
    Synonym Chlorotriethylstannane
    Chemical Name or Material Triethyltin chloride

    Specifications

    Formula Weight 241.33
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00040414
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Odor Unpleasant
    Sensitivity Moisture Sensitive
    Physical Form Liquid
    Synonym Chlorotriethylstannane
    Boiling Point 206°C
    TSCA TSCA
    CAS 994-31-0
    Quantity 25g
    UN Number UN2788
    Density 1.429g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
    Chemical Name or Material Triethyltin chloride

    GHS H Statement
    H300-H310-H330
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit