Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α,α-Trifluoro-m-tolunitrile, 96%, ACROS Organics™

Click to view available options
Quantity:
5mL
Packaging:
Glass bottle
368-77-4
C8H4F3N
171.122
MFCD00001807
OGOBINRVCUWLGN-UHFFFAOYSA-N
3-trifluoromethyl benzonitrile,3-cyanobenzotrifluoride,m-trifluoromethylbenzonitrile,alpha,alpha,alpha-trifluoro-m-tolunitrile,benzonitrile, 3-trifluoromethyl,a,a,a-trifluoro-m-tolunitrile,unii-y7zd7dk3ay,alpha,alpha,alpha-trifluoro-m-toluonitrile,y7zd7dk3ay,a,a,a-trifluor-m-tolunitril
67783
3-(trifluoromethyl)benzonitrile
C1=CC(=CC(=C1)C(F)(F)F)C#N
368-77-4
C8H4F3N
171.122
MFCD00001807
OGOBINRVCUWLGN-UHFFFAOYSA-N
3-trifluoromethyl benzonitrile,3-cyanobenzotrifluoride,m-trifluoromethylbenzonitrile,alpha,alpha,alpha-trifluoro-m-tolunitrile,benzonitrile, 3-trifluoromethyl,a,a,a-trifluoro-m-tolunitrile,unii-y7zd7dk3ay,alpha,alpha,alpha-trifluoro-m-toluonitrile,y7zd7dk3ay,a,a,a-trifluor-m-tolunitril
67783
3-(trifluoromethyl)benzonitrile
C1=CC(=CC(=C1)C(F)(F)F)C#N
Quantity 5mL
Color Colorless to Orange-Yellow
Density 1.2800g/mL
Infrared Spectrum Authentic
Refractive Index 1.4570 to 1.4610 (20°C, 589nm)
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula CF3C6H4CN
Beilstein 09, 478
Formula Weight 171.12
Physical Form Low Melting Solid
Percent Purity ≥95%
Chemical Name or Material α, α, α-Trifluoro-m-tolunitrile, 96%
Show More Show Less

GHS P Statement
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-711-4

TSCA : TSCA