Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(α,α,α-Trifluoro-p-tolyl)acetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
32857-62-8
C9H7F3O2
204.15
MFCD00004352
HNORVZDAANCHAY-UHFFFAOYSA-N
4-trifluoromethyl phenylacetic acid,2-4-trifluoromethyl phenyl acetic acid,4-trifluoromethylphenylacetic acid,4-trifluoromethyl phenyl acetic acid,alpha,alpha,alpha-trifluoro-p-tolyl acetic acid,benzeneacetic acid, 4-trifluoromethyl,p-trifluoromethyl phenylacetic acid,p-trifluoromethylphenylacetic acid,alpha,alpha,alpha-trifluoro-4-tolyl acetic acid
118342
2-[4-(trifluoromethyl)phenyl]acetic acid
C1=CC(=CC=C1CC(=O)O)C(F)(F)F
32857-62-8
C9H7F3O2
204.15
MFCD00004352
HNORVZDAANCHAY-UHFFFAOYSA-N
4-trifluoromethyl phenylacetic acid,2-4-trifluoromethyl phenyl acetic acid,4-trifluoromethylphenylacetic acid,4-trifluoromethyl phenyl acetic acid,alpha,alpha,alpha-trifluoro-p-tolyl acetic acid,benzeneacetic acid, 4-trifluoromethyl,p-trifluoromethyl phenylacetic acid,p-trifluoromethylphenylacetic acid,alpha,alpha,alpha-trifluoro-4-tolyl acetic acid
118342
2-[4-(trifluoromethyl)phenyl]acetic acid
C1=CC(=CC=C1CC(=O)O)C(F)(F)F
Infrared Spectrum Authentic
Melting Point 81°C to 85°C
Color White to Yellow
Packaging Glass bottle
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CF3C6H4CH2CO2H
Formula Weight 204.15
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material (α, α, α-Trifluoro-p-tolyl)acetic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 251-263-5