Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetaldehyde Ethyl Hemiacetal 90%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
433-27-2
C4H7F3O2
144.09
MFCD00000441
KLXJPQNHFFMLIG-UHFFFAOYSA-N
trifluoroacetaldehyde ethyl hemiacetal,fluoral ethyl hemiacetal,ethanol, 1-ethoxy-2,2,2-trifluoro,trifluoroacetaldehyde hemiethylacetal,1-ethoxy-2,2,2-trifluoro-ethanol,trifluoroacetaldehyde ethylhemiacetal,1-ethoxy-2,2,2-trifluoroethan-1-ol,pubchem12643,acmc-1aj38,1-ethoxy-2,2-trifluoroethanol
9897
1-ethoxy-2,2,2-trifluoroethanol
CCOC(C(F)(F)F)O
433-27-2
C4H7F3O2
144.09
MFCD00000441
KLXJPQNHFFMLIG-UHFFFAOYSA-N
trifluoroacetaldehyde ethyl hemiacetal,fluoral ethyl hemiacetal,ethanol, 1-ethoxy-2,2,2-trifluoro,trifluoroacetaldehyde hemiethylacetal,1-ethoxy-2,2,2-trifluoro-ethanol,trifluoroacetaldehyde ethylhemiacetal,1-ethoxy-2,2,2-trifluoroethan-1-ol,pubchem12643,acmc-1aj38,1-ethoxy-2,2-trifluoroethanol
9897
1-ethoxy-2,2,2-trifluoroethanol
CCOC(C(F)(F)F)O
Boiling Point 104°C to 105°C (745.0mmHg)
Flash Point 39°C
Specific Gravity 1.24
Refractive Index 1.3410 to 1.344
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.2400g/mL
Color Colorless
Quantity 25mL
Assay Percent Range 90%
Linear Formula CF3CH(OC2H5)OH
Beilstein 01, IV, 3133
Formula Weight 144.09
Physical Form Liquid
Percent Purity ≥88%
Chemical Name or Material Trifluoroacetaldehyde ethyl hemiacetal, 90%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 207-087-6

RTECSNumber : KK9000000

TSCA : TSCA