Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid 2,2,2-trifluoroethyl Ester 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
407-38-5
C4H2F6O2
196.048
MFCD00000418
ZKUJOCJJXCPCFS-UHFFFAOYSA-N
2,2,2-trifluoroethyl trifluoroacetate,acetic acid, trifluoro-, 2,2,2-trifluoroethyl ester,trifluoroacetic acid 2,2,2-trifluoroethyl ester,2,2,2-trifluoroethyltrifluoroacetate,pubchem12623,acmc-1ake7,cf3co2ch2cf3,cf3c o och2cf3,ksc496e0r,trifluoroethyl trifluoroacetate
67888
2,2,2-trifluoroethyl 2,2,2-trifluoroacetate
C(C(F)(F)F)OC(=O)C(F)(F)F
407-38-5
C4H2F6O2
196.048
MFCD00000418
ZKUJOCJJXCPCFS-UHFFFAOYSA-N
2,2,2-trifluoroethyl trifluoroacetate,acetic acid, trifluoro-, 2,2,2-trifluoroethyl ester,trifluoroacetic acid 2,2,2-trifluoroethyl ester,2,2,2-trifluoroethyltrifluoroacetate,pubchem12623,acmc-1ake7,cf3co2ch2cf3,cf3c o och2cf3,ksc496e0r,trifluoroethyl trifluoroacetate
67888
2,2,2-trifluoroethyl 2,2,2-trifluoroacetate
C(C(F)(F)F)OC(=O)C(F)(F)F
Color Colorless
Infrared Spectrum Authentic
Density 1.4640g/mL
Packaging Glass bottle
Quantity 25g
Assay Percent Range 98%
Linear Formula CF3CO2CH2CF3
Formula Weight 196.05
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Trifluoroacetic acid 2, 2, 2-trifluoroethyl ester
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger