Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
1kg
2.5kg
10kg
Packaging:
Glass Bottle
Plastic drum
76-05-1
C2HF3O2
114.023
MFCD00004169
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoracetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova
6422
CHEBI:45892
2,2,2-trifluoroacetic acid
C(=O)(C(F)(F)F)O
76-05-1
C2HF3O2
114.023
MFCD00004169
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid,perfluoroacetic acid,trifluoroethanoic acid,trifluoracetic acid,acetic acid, trifluoro,trifluoroaceticacid,cf3cooh,trifluoro acetic acid,trifluoro-acetic acid,kyselina trifluoroctova
6422
CHEBI:45892
2,2,2-trifluoroacetic acid
C(=O)(C(F)(F)F)O
Refractive Index 1.3
Water ≤0.3%
Melting Point -15.0°C
Infrared Spectrum Conforms to Reference
Boiling Point 72.0°C
Packaging Glass Bottle
Density 1.5350g/mL
Quantity 25g
Color Colorless
Vapor Pressure 107mbar at 25°C
Assay Percent Range 99%
Linear Formula CF3CO2H
Beilstein 02, II, 186
Fieser 01,1219; 02,433; 03,305; 04,530; 05,695; 06,613; 07,388; 08,503; 09,483; 10,418; 11,557; 12,529; 14,322; 15,338; 16,83; 17,340
Merck Index 15, 9847
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and hexane
Formula Weight 114.02
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Trifluoroacetic acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger