Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

76-05-1
C2HF3O2
114.023
DTQVDTLACAAQTR-UHFFFAOYSA-N
6422
CHEBI:45892

High quality chemicals for laboratory and specialized industrial use

CAS 76-05-1
Molecular Formula C2HF3O2
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES C(=O)(C(F)(F)F)O
IUPAC Name 2,2,2-trifluoroacetic acid
Molecular Weight (g/mol) 114.023
PubChem CID 6422
ChEBI CHEBI:45892
Formula Weight 114.03
Packaging Glass Bottle
Specific Gravity 1L = 1.53kg
Quantity 100mL
Chemical Name or Material Trifluoroacetic Acid
Grade BAKER ANALYZED™ Reagent
Show More Show Less