Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
10 x 1mL
500mL
70mL
900mL
Packaging:
Can
Glass Ampoule
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

76-05-1
C2HF3O2
114.023
DTQVDTLACAAQTR-UHFFFAOYSA-N
6422
CHEBI:45892
2,2,2-trifluoroacetic acid
C(=O)(C(F)(F)F)O

High purity solvents for use in analytical and preparative separations

Formula Weight 114.03
Packaging Can
Specific Gravity 1L = 1.53kg
Quantity 70mL
Chemical Name or Material Trifluoroacetic Acid, Gradient Tested for HPLC Peptide and Protein Analysis
Grade HPLC Grade