Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™

Catalog Number A1162AMP
Click to view available options
Quantity:
0.5 mL
1 mL
2 mL
10 mL
50 mL
Packaging:
Amber Glass
Ampule
Ampules
76-05-1
C2HF3O2
114.02
MFCD00004169
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
6422
CHEBI:45892
trifluoroacetic acid
OC(=O)C(F)(F)F
76-05-1
C2HF3O2
114.02
MFCD00004169
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
6422
CHEBI:45892
trifluoroacetic acid
OC(=O)C(F)(F)F

Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL

  • Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
  • Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
  • Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY
Absorbance 0.003 AU max. at 254nm, 0.03 AU max. at 300nm, 0.06 AU max. at 230nm, 0.06 AU max. at 275nm, 0.18 AU max. at 220nm, 0.55 AU max. at 210nm
pH 0.45
Melting Point -15°C
Boiling Point 72°C
Color Colorless
Assay Percent Range ≥99.5 %
Linear Formula CF3COOH
Merck Index 15, 9847
Formula Weight 114.02
Percent Purity ≥99.5%
Grade OPTIMA™ LC-MS
Density 1.535g/cm³
Quantity 2 mL
Packaging Ampule
Physical Form Liquid
Residue after Evaporation 1ppm max.
Vapor Density 3.9
Vapor Pressure 107mbar 25
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe burns by all exposure routes. Inhalation may cause central nervous system effects. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit