Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic anhydride, +99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
250g
500g
2.5kg
Packaging:
Glass Bottle
407-25-0
C4F6O3
210.031
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
trifluoroacetic anhydride,trifluoroacetic acid anhydride,2,2,2-trifluoroacetic anhydride,bis trifluoroacetic anhydride,acetic acid, trifluoro-, anhydride,perfluoroacetic anhydride,trifluoroacetyl anhydride,tfaa,hexafluoroacetic anhydride,anhydrid kyseliny trifluoroctove
9845
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
C(=O)(C(F)(F)F)OC(=O)C(F)(F)F
407-25-0
C4F6O3
210.031
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
trifluoroacetic anhydride,trifluoroacetic acid anhydride,2,2,2-trifluoroacetic anhydride,bis trifluoroacetic anhydride,acetic acid, trifluoro-, anhydride,perfluoroacetic anhydride,trifluoroacetyl anhydride,tfaa,hexafluoroacetic anhydride,anhydrid kyseliny trifluoroctove
9845
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
C(=O)(C(F)(F)F)OC(=O)C(F)(F)F
Acylation reagent
Melting Point -63.5°C
Quantity 25g
Vapor Pressure 141mbar at 20°C
Packaging Glass Bottle
Refractive Index 1.3
Infrared Spectrum Authentic
Color Undesignated
Density 1.5100g/mL
Boiling Point 39.1°C
Assay Percent Range 99+%
Linear Formula (CF3CO)2O
Beilstein 02, II, 186
Fieser 01,1221; 03,308; 05,701; 06,616; 07,389; 09,484; 12,530; 14,151; 15,14; 01,373
Solubility Information Solubility in water: hydrolysis. Other solubilities: miscible with polar organic solvents, partically soluble in unpolar hydrocarbons
Formula Weight 210.03
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Trifluoroacetic anhydride
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger