Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetylacetone 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
367-57-7
C5H5F3O2
154.088
MFCD00000427
SHXHPUAKLCCLDV-UHFFFAOYSA-N
1,1,1-trifluoro-2,4-pentanedione,trifluoroacetylacetone,1,1,1-trifluoroacetylacetone,trifluoroacetyl acetone,acetyl trifluoroacetone,2,4-pentanedione, 1,1,1-trifluoro,tfpd-h,unii-9n20a8g8sw,alpha,alpha,alpha-trifluoroacetylacetone,a,a,a-trifluoroacetylacetone
73943
1,1,1-trifluoropentane-2,4-dione
CC(=O)CC(=O)C(F)(F)F
367-57-7
C5H5F3O2
154.088
MFCD00000427
SHXHPUAKLCCLDV-UHFFFAOYSA-N
1,1,1-trifluoro-2,4-pentanedione,trifluoroacetylacetone,1,1,1-trifluoroacetylacetone,trifluoroacetyl acetone,acetyl trifluoroacetone,2,4-pentanedione, 1,1,1-trifluoro,tfpd-h,unii-9n20a8g8sw,alpha,alpha,alpha-trifluoroacetylacetone,a,a,a-trifluoroacetylacetone
73943
1,1,1-trifluoropentane-2,4-dione
CC(=O)CC(=O)C(F)(F)F
Color Yellow
Boiling Point 101°C
Quantity 100g
UN Number 1224
Formula Weight 154.09
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Trifluoroacetylacetone