Learn More
Trifluoromethanesulfonamide, 96%, Thermo Scientific Chemicals
Description
Trifluoromethanesulfonamide is used as an intermediate in synthetic chemistry. It is used in the preparation of specific inhibitors of mammalian secreted phospholipases A2. It is also involved in the synthesis of ecicosanoids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trifluoromethanesulfonamide is used as an intermediate in synthetic chemistry. It is used in the preparation of specific inhibitors of mammalian secreted phospholipases A2. It is also involved in the synthesis of ecicosanoids.
Solubility
Soluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
Specifications
Melting Point | 116°C to 119°C |
Quantity | 5 g |
Beilstein | 1812099 |
Solubility Information | Soluble in water. |
Formula Weight | 149.09 |
Percent Purity | 96% |
Chemical Name or Material | Trifluoromethanesulfonamide |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 431-270-1 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.