Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trifluoromethanesulfonamide, 96%

Click to view available options
Quantity:
1g
5g
421-85-2
CH2F3NO2S
149.087
MFCD00068714
KAKQVSNHTBLJCH-UHFFFAOYSA-N
trifluoromethanesulphonamide,trifluoromethane sulfonamide,trifluoromethylsulfonamide,1,1,1-trifluoromethanesulfonamide,chembl67511,methanesulfonamide, 1,1,1-trifluoro,1bcd,trifluoromethyl sulfonamide,triflamide,trifluorometanesulfonamide
79001
trifluoromethanesulfonamide
C(F)(F)(F)S(=O)(=O)N
421-85-2
CH2F3NO2S
149.087
MFCD00068714
KAKQVSNHTBLJCH-UHFFFAOYSA-N
trifluoromethanesulphonamide,trifluoromethane sulfonamide,trifluoromethylsulfonamide,1,1,1-trifluoromethanesulfonamide,chembl67511,methanesulfonamide, 1,1,1-trifluoro,1bcd,trifluoromethyl sulfonamide,triflamide,trifluorometanesulfonamide
79001
trifluoromethanesulfonamide
C(F)(F)(F)S(=O)(=O)N
Applications
Trifluoromethanesulfonamide is used as an intermediate in synthetic chemistry. It is used in the preparation of specific inhibitors of mammalian secreted phospholipases A2. It is also involved in the synthesis of ecicosanoids.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
Melting Point 116°C to 119°C
Quantity 1g
Beilstein 1812099
Formula Weight 149.09
Percent Purity 96%
Chemical Name or Material Trifluoromethanesulfonamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning