Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoromethanesulfonic anhydride, 98+%, ACROS Organics™

Click to view available options
Quantity:
1mL
50mL
250mL
Packaging:
Glass bottle
358-23-6
C2F6O5S2
282.13
MFCD00000408
WJKHJLXJJJATHN-UHFFFAOYSA-N
trifluoromethanesulfonic anhydride,triflic anhydride,trifluoromethanesulphonic anhydride,trifluoromethanesulfonic acid anhydride,methanesulfonic acid, trifluoro-, anhydride,trifluoromethanesulphonic acid anhydride,unii-8w034lhg1u,tf2o,trifluoromethane sulfonyl trifluoromethanesulfonate,trifluoromethane sulfonic anhydride
67749
CHEBI:48509
trifluoromethylsulfonyl trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F
358-23-6
C2F6O5S2
282.13
MFCD00000408
WJKHJLXJJJATHN-UHFFFAOYSA-N
trifluoromethanesulfonic anhydride,triflic anhydride,trifluoromethanesulphonic anhydride,trifluoromethanesulfonic acid anhydride,methanesulfonic acid, trifluoro-, anhydride,trifluoromethanesulphonic acid anhydride,unii-8w034lhg1u,tf2o,trifluoromethane sulfonyl trifluoromethanesulfonate,trifluoromethane sulfonic anhydride
67749
CHEBI:48509
trifluoromethylsulfonyl trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F

Trifluoromethanesulfonic anhydride, 98+%, ACROS Organics, Quantity: 250mL, Packaging: Glass bottle, Boiling Point: 80.0 deg.C, Colorless to Brown, Molecular Weight: 282.13, Percent Purity: 98+%, Assay Percent Range: 98+%, Beilstein: 03, IV, 35, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.7000g/mL

Trifluoromethanesulfonic anhydride, 98+%, ACROS Organics, Quantity: 250mL, Packaging: Glass bottle, Boiling Point: 80.0 deg.C, Colorless to Brown, Molecular Weight: 282.13, Percent Purity: 98+%, Assay Percent Range: 98+%, Beilstein: 03, IV, 35, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.7000g/mL

Alkylation reagent, Coupling reagent
Specific Gravity 1.7
Refractive Index 1.3200 to 1.3230
Color Colorless to Brown
Infrared Spectrum Authentic
Quantity 50mL
Density 1.7000g/mL
Packaging Glass bottle
Boiling Point 80.0°C
Assay Percent Range 98+%
Linear Formula (CF3SO2)2O
Beilstein 03, IV, 35
Fieser 04,533; 05,702; 06,618; 07,390; 10,419; 11,560; 12,532; 14,324; 15,339; 16,357; 17,156
Merck Index 15, 9850
Solubility Information Solubility in water: reacts viontly with water
Formula Weight 282.13
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Trifluoromethanesulfonic anhydride
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
May be corrosive to metals.

GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

GHS Signal Word: Danger

EINECSNumber : 206-616-8