Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trigonelline hydrochloride, MilliporeSigma™ Supelco™

$38.90 - $260.50

Specifications

CAS 6138-41-6
Grade Analytical Standard
Linear Formula C7H7 NO2 · HCl
Physical Form Neat
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4893
View Documents
MilliporeSigma Supelco
T55091G
1 g
Each for $38.90
Add to cart
 
11-101-4894
View Documents
MilliporeSigma Supelco
T55095G
5 g
Each for $136.10
Add to cart
 
11-101-4895
View Documents
MilliporeSigma Supelco
T550910G
10 g
Each for $260.50
Add to cart
 
Description

Description

Featured Industries: Food and Beverages
Specifications

Specifications

6138-41-6
Analytical Standard
C7H7 NO2 · HCl
Neat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit