Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triisopropyl orthoformate, 97%

Click to view available options
Quantity:
25g
100g
4447-60-3
C10H22O3
190.283
MFCD00060628
FPIVAWNGRDHRSQ-UHFFFAOYSA-N
triisopropyl orthoformate,triisopropoxymethane,triisopropylorthoformate,propane, 2,2',2-methylidynetris oxy tris,2-diisopropoxymethoxy propane,2-di propan-2-yloxy methoxy propane,2-bis propan-2-yloxy methoxy propane,orthoformic acid, triisopropyl ester,isopropyl orthoformate
78191
2-[di(propan-2-yloxy)methoxy]propane
CC(C)OC(OC(C)C)OC(C)C
4447-60-3
C10H22O3
190.283
MFCD00060628
FPIVAWNGRDHRSQ-UHFFFAOYSA-N
triisopropyl orthoformate,triisopropoxymethane,triisopropylorthoformate,propane, 2,2',2-methylidynetris oxy tris,2-diisopropoxymethoxy propane,2-di propan-2-yloxy methoxy propane,2-bis propan-2-yloxy methoxy propane,orthoformic acid, triisopropyl ester,isopropyl orthoformate
78191
2-[di(propan-2-yloxy)methoxy]propane
CC(C)OC(OC(C)C)OC(C)C
Odor Pungent
Flash Point 42°C (108°F)
Quantity 25g
Boiling Point 64°C to 65°C (18mmHg)
UN Number UN3272
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material Triisopropyl orthoformate

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA