Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Triisopropyl orthoformate, 97%

Click to view available options
Quantity:
25g
100g
4447-60-3
C10H22O3
190.283
MFCD00060628
FPIVAWNGRDHRSQ-UHFFFAOYSA-N
triisopropyl orthoformate,triisopropoxymethane,triisopropylorthoformate,propane, 2,2',2-methylidynetris oxy tris,2-diisopropoxymethoxy propane,2-di propan-2-yloxy methoxy propane,2-bis propan-2-yloxy methoxy propane,orthoformic acid, triisopropyl ester,isopropyl orthoformate
78191
2-[di(propan-2-yloxy)methoxy]propane
CC(C)OC(OC(C)C)OC(C)C
4447-60-3
C10H22O3
190.283
MFCD00060628
FPIVAWNGRDHRSQ-UHFFFAOYSA-N
triisopropyl orthoformate,triisopropoxymethane,triisopropylorthoformate,propane, 2,2',2-methylidynetris oxy tris,2-diisopropoxymethoxy propane,2-di propan-2-yloxy methoxy propane,2-bis propan-2-yloxy methoxy propane,orthoformic acid, triisopropyl ester,isopropyl orthoformate
78191
2-[di(propan-2-yloxy)methoxy]propane
CC(C)OC(OC(C)C)OC(C)C
Sensitivity Moisture sensitive
Density 0.854
Refractive Index 1.397
Boiling Point 64°C to 65°C (18mmHg)
Quantity 100g
Flash Point 42°C (108°F)
Odor Pungent
UN Number UN3272
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material Triisopropyl orthoformate
Show More Show Less

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 224-688-9

TSCA : Yes

Recommended Storage : Ambient temperatures