Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropyl phosphite, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
116-17-6
C9H21O3P
208.24
SJHCUXCOGGKFAI-UHFFFAOYSA-N
triisopropyl phosphite,tri-2-propyl phosphite,phosphorous acid, tris 1-methylethyl ester,isopropyl phosphite, tri,triisopropoxyphosphine,phosphorous acid, triisopropyl ester,tri-2-propylphosphite,unii-rj7qu4yqun,triisopropylphosphite,tri-i-propylphosphite
8304
tripropan-2-yl phosphite
CC(C)OP(OC(C)C)OC(C)C
116-17-6
C9H21O3P
208.24
SJHCUXCOGGKFAI-UHFFFAOYSA-N
triisopropyl phosphite,tri-2-propyl phosphite,phosphorous acid, tris 1-methylethyl ester,isopropyl phosphite, tri,triisopropoxyphosphine,phosphorous acid, triisopropyl ester,tri-2-propylphosphite,unii-rj7qu4yqun,triisopropylphosphite,tri-i-propylphosphite
8304
tripropan-2-yl phosphite
CC(C)OP(OC(C)C)OC(C)C
Boiling Point 94°C to 95°C (66.6 hPa)
Specific Gravity 0.914
Color Undesignated
Density 0.9140g/mL
Infrared Spectrum Authentic
Flash Point 62°C
Refractive Index 1.4095 to 1.4115
Quantity 5g
Packaging Glass bottle
Assay Percent Range 95% min. (GC)
Linear Formula [(CH3)2CHO]3P
Beilstein 01,363
Fieser 01,1229; 16,36
Solubility Information Solubility in water: insoluble.
Formula Weight 208.24
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Triisopropyl phosphite
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 204-130-