Alfa Aesar™ Triisopropylphosphine, 98%

Manufacturer: Alfa Aesar™ 4168402

Quantity 0.5g

 View more versions of this product

Catalog No. AA4168402

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6476-36-4
    Formula Weight 160.24
    MDL Number MFCD00015029
    UN Number UN2845
    Chemical Name or Material Triisopropylphosphine

    Specifications

    Formula Weight 160.24
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.4670
    MDL Number MFCD00015029
    CAS 6476-36-4
    UN Number UN2845
    Odor Pungent
    Sensitivity Air Sensitive
    Boiling Point 176° to 178°C
    Physical Form Liquid
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Density 0.839g/mL
    Flash Point −17° (1°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H250-H252-H314-H318
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 0.5g
    Chemical Name or Material Triisopropylphosphine
    Assay 98%

    GHS H Statement
    H250-H252-H314-H318
    Catches fire spontaneously if exposed to air.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit