Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisopropylsilane, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
6485-79-6
C9H22Si
158.36
MFCD00009657
ZGYICYBLPGRURT-UHFFFAOYSA-N
triisopropylsilane,tris propan-2-yl silane,triisopropyl silane,tri propan-2-yl silicon,tri-isopropylsilyl radical,triisopropylsilyl,tri-iso-propylsilane,ambotzrl-1102,pubchem12855
6327611
tri(propan-2-yl)silicon
CC(C)[Si](C(C)C)C(C)C
6485-79-6
C9H22Si
158.36
MFCD00009657
ZGYICYBLPGRURT-UHFFFAOYSA-N
triisopropylsilane,tris propan-2-yl silane,triisopropyl silane,tri propan-2-yl silicon,tri-isopropylsilyl radical,triisopropylsilyl,tri-iso-propylsilane,ambotzrl-1102,pubchem12855
6327611
tri(propan-2-yl)silicon
CC(C)[Si](C(C)C)C(C)C
Reduction reagent
Specific Gravity 0.773
Flash Point 37°C
Quantity 50g
Refractive Index 1.4340 to 1.4360
Boiling Point 84.0°C to 86.0°C (35.0mmHg)
Density 0.7730g/mL
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Linear Formula [(CH3)2CH]3SiH
Beilstein 04,IV,3907
Solubility Information Solubility in water: negligible
Formula Weight 158.36
Percent Purity 98%
Chemical Name or Material Triisopropylsilane, 98%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning