Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Triisopropylsilyl)acetylene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
89343-06-6
C11H22Si
182.38
MFCD00075452
KZGWPHUWNWRTEP-UHFFFAOYSA-N
triisopropylsilyl acetylene,ethynyltriisopropylsilane,triisopropylsilylacetylene,ethynyltris propan-2-yl silane,ethynyl-tri propan-2-yl silane,silane, ethynyltris 1-methylethyl,triisopropylsilylacetylen,acmc-209qzk,ethynyltriisopropyl-silane
2734682
ethynyl-tri(propan-2-yl)silane
CC(C)[Si](C#C)(C(C)C)C(C)C
89343-06-6
C11H22Si
182.38
MFCD00075452
KZGWPHUWNWRTEP-UHFFFAOYSA-N
triisopropylsilyl acetylene,ethynyltriisopropylsilane,triisopropylsilylacetylene,ethynyltris propan-2-yl silane,ethynyl-tri propan-2-yl silane,silane, ethynyltris 1-methylethyl,triisopropylsilylacetylen,acmc-209qzk,ethynyltriisopropyl-silane
2734682
ethynyl-tri(propan-2-yl)silane
CC(C)[Si](C#C)(C(C)C)C(C)C
Boiling Point 50°C to 52°C (6.0mmHg)
Flash Point 56°C
Specific Gravity 0.813
Density 0.8130g/mL
Refractive Index 1.4515 to 1.4535
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula [(CH3)2CH]3SiC∽CH
Formula Weight 182.38
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material (Triisopropylsilyl)acetylene, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning