Alfa Aesar™ Triisopropylsilylacetylene, 97%

Manufacturer: Alfa Aesar™ H5340514

Quantity 25g

 View more versions of this product

Catalog No. AAH5340514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89343-06-6
    Molecular Formula C11H22Si
    Linear Formula [(CH3)2CH]3SiC≡CH
    Formula Weight 182.38
    MDL Number MFCD00075452
    UN Number UN1993
    Beilstein 3536241
    Synonym Ethynyltriisopropylsilane
    Chemical Name or Material Triisopropylsilylacetylene

    Specifications

    Formula Weight 182.38
    Quantity 25g
    Flash Point 56°C (133°F)
    Molecular Formula C11H22Si
    Linear Formula [(CH3)2CH]3SiC≡CH
    MDL Number MFCD00075452
    UN Number UN1993
    Synonym Ethynyltriisopropylsilane
    Boiling Point 50° to 52°C (0.6mmHg)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 89343-06-6
    Beilstein 3536241
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Triisopropylsilylacetylene
    Assay 97%

    Description

    Applications
    Triisopropylsilylacetylene is used as a reagent for the rhodium-catalyzed asymmetric alkylation of alpha, beta-unsaturated carbonyl compounds as well as in the synthesis of beta-alkynylated nitroalkanes. It is utilized in the preparation of bromoethynyl-triisopropyl-silane in the presence of the reagents such as N-bromo succinimide and silver nitrate. It is actively in involved in Sonogashira coupling reaction with 1-bromo-3-iodo-5-tertbutylbenzene.

    Solubility
    Miscible with organic solvents.

    Notes
    Incompatible with strong acids, strong bases and strong oxidizing agents.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit