Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110516
Description
Immunogen sequence: VCRVPGQIEV LRGFLEDVVP DATSAYPYLL LYESRQRRYL GSSPEGSGFC SKDRFVAYPC AVGQTAFSSG RHYWEVGMNI TGDALWALGV CRDNVSRKDR VPKCPENGFW VVQLSKGTKY LSTFSALTPV MLMEPPSHMG IFLDFEAGEV SFYSVSDGSH LHTYSQATFP GPLQPFFCLG APKSGQMVIS TVTMWVKG.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
TRIM17 | |
Polyclonal | |
Unconjugated | |
TRIM17 | |
E3 ubiquitin-protein ligase TRIM17; RBCC; RING finger protein 16; RING finger protein terf; RING-type E3 ubiquitin transferase TRIM17; RNF16; terf; Testis RING finger protein; Trim17; tripartite motif containing 17; tripartite motif protein 17; tripartite motif protein TRIM17; tripartite motif-containing 17; tripartite motif-containing protein 17; Unknown (protein for MGC:134518) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51127, 56631, 64702 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q7TPM3, Q9WV59, Q9Y577 | |
TRIM17 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 280-477 of human TRIM17 (NP_0571861). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction