Learn More
TRIM24 Mouse anti-Human, Clone: OTI1D4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802795
Catalog No. 50-166-6166
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
Specifications
TRIM24
Monoclonal
Unconjugated
TRIM24
Lyophilized
TRIM24
Human recombit protein fragment corresponding to amino acids 706-1016 of human TRIM24 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1D4
PBS with 8% trehalose and no preservative; pH 7.3
O15164
PTC6, RNF82, TF1A, TIF1, TIF1A, TIF1ALPHA, hTIF1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8805