Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM24 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
Supplier: Active Motif 61551

Description
The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
TRIM24 | |
Polyclonal | |
Unconjugated | |
TRIM24 | |
A130082H20Rik; Ab2-427; AI447469; D430004I05Rik; E3 ubiquitin-protein ligase Trim24; hTIF1; PTC6; RING finger protein 82; RING-type E3 ubiquitin transferase TIF1-alpha; RNF82; TF1A; Tif1; TIF1A; TIF1alpha; TIF1-alpha; Transcription intermediary factor 1-alpha; transcriptional intermediary factor 1; transcriptional intermediary factor 1, alpha; TRIM24; tripartite motif containing 24; tripartite motif-containing 24; Tripartite motif-containing protein 24 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8805 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
Lot-Specific | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
O15164 | |
TRIM24 | |
This antibody was raised against a peptide within the C-terminal region of human TRIM24. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title