Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM25 Antibody (SR2171), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | TRIM25 |
---|---|
Clone | SR2171 |
Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:60, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
TRIM25 Monoclonal antibody specifically detects TRIM25 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
TRIM25 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:60, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 6.3.2.19, EC 6.3.2.n3, EFPZ147, Estrogen-responsive finger protein, RING finger protein 147, RNF147E3 ubiquitin/ISG15 ligase TRIM25, tripartite motif containing 25, tripartite motif-containing 25, Tripartite motif-containing protein 25, Ubiquitin/ISG15-conjugating enzyme TRIM25, Zinc finger protein 147, zinc finger protein 147 (estrogen-responsive finger protein), zinc finger protein-147, ZNF147tripartite motif protein TRIM25 | |
A synthesized peptide derived from human TRIM25 (Uniprot #: Q14258) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2171 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Zinc Finger | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
7706 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title